Please login to your online account to display your discounted pricing

4-Benzyl-2-(chloromethyl)morpholine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-Benzyl-2-(chloromethyl)morpholine
CAS 40987-25-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H16ClNO
Formula Weight 225.72
MDL Number MFCD02681886
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL01000EA

 
fisher scientific company
TL01000EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.72
Quantity 10g
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material 4-Benzyl-2-(chloromethyl)morpholine
MDL Number MFCD02681886
CAS 40987-25-5
Packaging Amber Glass Bottle
Molecular Formula C12H16ClNO
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.