Please login to your online account to display your discounted pricing

4-Benzyl-2-(chloromethyl)morpholine, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  TL01000EA

 View more versions of this product

Catalog No. TL01000EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 225.72
Quantity 10g
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material 4-Benzyl-2-(chloromethyl)morpholine
MDL Number MFCD02681886
CAS 40987-25-5
Packaging Amber Glass Bottle
Molecular Formula C12H16ClNO
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Benzyl-2-(chloromethyl)morpholine
CAS 40987-25-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H16ClNO
Formula Weight 225.72
MDL Number MFCD02681886
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents