missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzoylpyridine, 98%, ACROS Organics™

$116.45 - $116.45

Chemical Identifiers

CAS 14548-46-0
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD00006430
InChI Key SKFLCXNDKRUHTA-UHFFFAOYSA-N
Synonym 4-benzoyl pyridine, 4-benzoylpyridine, 4-pyridyl phenyl ketone, gamma-benzoylpyridine, ketone, phenyl 4-pyridyl, methanone, phenyl-4-pyridinyl, phenyl 4-pyridinyl methanone, phenyl 4-pyridyl ketone, phenyl pyridin-4-yl methanone, pyridine, 4-benzoyl
PubChem CID 26731
IUPAC Name phenyl(pyridin-4-yl)methanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=NC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC154090050
SDS View Documents
Thermo Fisher Scientific
154090050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC154091000
SDS View Documents
Thermo Fisher Scientific
154091000
100g Plastic bottle
Each for $116.45
 
Description

Description

Specifications

Chemical Identifiers

14548-46-0
183.21
SKFLCXNDKRUHTA-UHFFFAOYSA-N
26731
C1=CC=C(C=C1)C(=O)C2=CC=NC=C2
C12H9NO
MFCD00006430
4-benzoyl pyridine, 4-benzoylpyridine, 4-pyridyl phenyl ketone, gamma-benzoylpyridine, ketone, phenyl 4-pyridyl, methanone, phenyl-4-pyridinyl, phenyl 4-pyridinyl methanone, phenyl 4-pyridyl ketone, phenyl pyridin-4-yl methanone, pyridine, 4-benzoyl
phenyl(pyridin-4-yl)methanone

Specifications

150°C
Glass bottle
Yellow
5g
97.5
97.5% min. (GC)
C6H5CO(C5H4N)
21, 331
Solubility in water: insoluble. Other solubilities: soluble in methanol,very soluble in ether & ethanol
C1=CC=C(C=C1)C(=O)C2=CC=NC=C2
183.21
183.21
98%
Authentic
315.0°C
68.0°C to 72.0°C
14548-46-0
100.0
C12H9NO
MFCD00006430
4-benzoyl pyridine, 4-benzoylpyridine, 4-pyridyl phenyl ketone, gamma-benzoylpyridine, ketone, phenyl 4-pyridyl, methanone, phenyl-4-pyridinyl, phenyl 4-pyridinyl methanone, phenyl 4-pyridyl ketone, phenyl pyridin-4-yl methanone, pyridine, 4-benzoyl
SKFLCXNDKRUHTA-UHFFFAOYSA-N
phenyl(pyridin-4-yl)methanone
26731
Crystalline Powder or Crystalline Chunks
4-Benzoylpyridine, 98%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Warning

EINECSNumber : 238-586-7

RTECSNumber : OB6402000

TSCA : TSCA

SDS