Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzoylpyridine, 97%

$45.09 - $155.47

Chemical Identifiers

Chemical Name or Material 4-Benzoylpyridine
CAS 14548-46-0
Molecular Formula C12H9NO
Formula Weight 183.21
MDL Number MFCD00006430
Beilstein 3864
Synonym Phenyl 4-pyridyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1794530

 
alfa aesar™
A1794530
250g Each for $155.47

AAA1794518

 
alfa aesar™
A1794518
50g Each for $45.09
Description & Specifications

Specifications

Formula Weight 183.21
Chemical Name or Material 4-Benzoylpyridine
TSCA TSCA
Assay 97%
Molecular Formula C12H9NO
MDL Number MFCD00006430
CAS 14548-46-0
Beilstein 3864
Synonym Phenyl 4-pyridyl ketone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 130° to 131°C (1mmHg)
Melting Point 69° to 71°C
Quantity 250g
Flash Point 150°C (302°F)
RTECS Number OB6402000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.