missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzoylpiperidine hydrochloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 232860010

 View more versions of this product

Catalog No. AC232860010



Specifications

Specifications

4-Benzoylpiperidine hydrochloride
226°C to 227°C
98%
97.5
98%
MFCD00066982
Solubility in water: soluble in water. Other solubilities: soluble in methanol and ethanol
C1CNCCC1C(=O)C2=CC=CC=C2.Cl
225.72
225.72
Glass bottle
1g
25519-80-6
100.0
C12H15NO·ClH
4-benzoyl piperidine hcl, 4-benzoyl piperidine hydrochloride, 4-benzoylpiperidine hcl, 4-benzoylpiperidine hydrochloride, 4-benzoylpiperidine hydrogen chloride, methanone, phenyl-4-piperidinyl-, hydrochloride, phenyl 4-piperidyl ketone, chloride, phenyl 4-piperidyl methanone hydrochloride, phenyl piperidin-4-yl methanone hydrochloride, pubchem9881
NXYKIFZJQXOUJS-UHFFFAOYSA-N
phenyl(piperidin-4-yl)methanone;hydrochloride
2724437
98%

Chemical Identifiers

25519-80-6
225.72
NXYKIFZJQXOUJS-UHFFFAOYSA-N
2724437
C1CNCCC1C(=O)C2=CC=CC=C2.Cl
C12H15NO·ClH
MFCD00066982
4-benzoyl piperidine hcl, 4-benzoyl piperidine hydrochloride, 4-benzoylpiperidine hcl, 4-benzoylpiperidine hydrochloride, 4-benzoylpiperidine hydrogen chloride, methanone, phenyl-4-piperidinyl-, hydrochloride, phenyl 4-piperidyl ketone, chloride, phenyl 4-piperidyl methanone hydrochloride, phenyl piperidin-4-yl methanone hydrochloride, pubchem9881
phenyl(piperidin-4-yl)methanone;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents