missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzoylbutyric acid, 97%, ACROS Organics™

$203.00 - $203.00

Chemical Identifiers

CAS 1501-05-9
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.21
MDL Number MFCD00004411
InChI Key SHKWSBAVRQZYLE-UHFFFAOYSA-N
Synonym 4-benzoyl-butyric acid
PubChem CID 73914
IUPAC Name 5-oxo-5-phenylpentanoic acid
SMILES C1=CC=C(C=C1)C(=O)CCCC(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC105740250 SDS View Documents Thermo Fisher Scientific
AC105740250
25g Glass bottle Each for $203.00
 
Specifications

Chemical Identifiers

1501-05-9
192.21
SHKWSBAVRQZYLE-UHFFFAOYSA-N
73914
C1=CC=C(C=C1)C(=O)CCCC(=O)O
C11H12O3
MFCD00004411
4-benzoyl-butyric acid
5-oxo-5-phenylpentanoic acid

Specifications

Authentic
White to Beige
25g
96.0
97%
C6H5CO(CH2)3CO2H
10, 708
Solubility in water: insoluble in water. Other solubilities: soluble in benzene (warm),chloroform,ethanol,,dichloromethane,ethyl acetate,aqueous alkali,insoluble in hexane
C1=CC=C(C=C1)C(=O)CCCC(=O)O
192.21
192.21
97%
Glass bottle
124°C to 128°C
1501-05-9
100.0
C11H12O3
MFCD00004411
4-benzoyl-butyric acid
SHKWSBAVRQZYLE-UHFFFAOYSA-N
5-oxo-5-phenylpentanoic acid
73914
Crystalline Powder
4-Benzoylbutyric acid, 97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-113-5