Learn More
4-Benzoylbutyric acid, 97%, Thermo Scientific Chemicals
$88.90 - $301.73
Chemical Identifiers
CAS | 1501-05-9 |
---|---|
Molecular Formula | C11H12O3 |
Molecular Weight (g/mol) | 192.214 |
MDL Number | MFCD00004411 |
InChI Key | SHKWSBAVRQZYLE-UHFFFAOYSA-N |
Synonym | 4-benzoylbutyric acid, 5-oxo-5-phenylvaleric acid, benzenepentanoic acid, .delta.-oxo, 4-benzoyl-butyric acid, timtec-bb sbb007811, 5-oxo-5-phenyl-pentanoic acid, 4-benzoylbutyric acid;4-benzoylbutanoic acid, butanoic acid, 4-benzoyl, 4-benzoylbutyricacid |
PubChem CID | 73914 |
IUPAC Name | 5-oxo-5-phenylpentanoic acid |
SMILES | C1=CC=C(C=C1)C(=O)CCCC(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0401306
|
Thermo Scientific Chemicals
L0401306 |
5 g |
Each for $88.90
|
|
|||||
AAL0401314
|
Thermo Scientific Chemicals
L0401314 |
25 g |
Each for $301.73
|
|
|||||
Description
4-Benzoylbutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Benzoylbutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
1501-05-9 | |
192.214 | |
SHKWSBAVRQZYLE-UHFFFAOYSA-N | |
73914 | |
C1=CC=C(C=C1)C(=O)CCCC(=O)O |
C11H12O3 | |
MFCD00004411 | |
4-benzoylbutyric acid, 5-oxo-5-phenylvaleric acid, benzenepentanoic acid, .delta.-oxo, 4-benzoyl-butyric acid, timtec-bb sbb007811, 5-oxo-5-phenyl-pentanoic acid, 4-benzoylbutyric acid;4-benzoylbutanoic acid, butanoic acid, 4-benzoyl, 4-benzoylbutyricacid | |
5-oxo-5-phenylpentanoic acid |
Specifications
1501-05-9 | |
C11H12O3 | |
5 g | |
4-benzoylbutyric acid, 5-oxo-5-phenylvaleric acid, benzenepentanoic acid, .delta.-oxo, 4-benzoyl-butyric acid, timtec-bb sbb007811, 5-oxo-5-phenyl-pentanoic acid, 4-benzoylbutyric acid;4-benzoylbutanoic acid, butanoic acid, 4-benzoyl, 4-benzoylbutyricacid | |
SHKWSBAVRQZYLE-UHFFFAOYSA-N | |
5-oxo-5-phenylpentanoic acid | |
73914 | |
97% |
126°C to 128°C | |
MFCD00004411 | |
641947 | |
Insoluble in water. | |
C1=CC=C(C=C1)C(=O)CCCC(=O)O | |
192.214 | |
192.21 | |
4-Benzoylbutyric acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 216-113-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only