missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzoylbiphenyl, 99%

Manufacturer:  Alfa Aesar™ A1557336

 View more versions of this product

Catalog No. AAA1557336



Specifications

Specifications

4-Benzoylbiphenyl
500g
C19H14O
1876092
LYXOWKPVTCPORE-UHFFFAOYSA-N
phenyl-(4-phenylphenyl)methanone
75040
99%
98°C to 103°C
2128-93-0
MFCD00003079
4-benzoylbiphenyl, 4-phenylbenzophenone, p-phenylbenzophenone, benzophenone, 4-phenyl, p-benzoylbiphenyl, 4-diphenyl phenyl ketone, p-biphenylyl phenyl ketone, trigonal 12, 4-biphenylyl phenyl ketone, methanone, 1,1'-biphenyl-4-ylphenyl
C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C3=CC=CC=C3
258.32
258.32

Chemical Identifiers

2128-93-0
258.32
LYXOWKPVTCPORE-UHFFFAOYSA-N
75040
C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C3=CC=CC=C3
C19H14O
MFCD00003079
4-benzoylbiphenyl, 4-phenylbenzophenone, p-phenylbenzophenone, benzophenone, 4-phenyl, p-benzoylbiphenyl, 4-diphenyl phenyl ketone, p-biphenylyl phenyl ketone, trigonal 12, 4-biphenylyl phenyl ketone, methanone, 1,1'-biphenyl-4-ylphenyl
phenyl-(4-phenylphenyl)methanone
Safety and Handling

Safety and Handling

RTECSNumber : PC4936800

TSCA : TSCA

Documents