missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzoylbenzoic Acid, TCI America™

$125.28 - $380.42

Chemical Identifiers

CAS 611-95-0
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.231
MDL Number MFCD00002560
InChI Key IFQUPKAISSPFTE-UHFFFAOYSA-N
Synonym p-benzoylbenzoic acid, benzoic acid, 4-benzoyl, 4-carboxybenzophenone, 4-benzoyl-benzoic acid, 4-phenylcarbonyl benzoic acid, benzoic acid, p-benzoyl, benzophenone-4-carboxylic acid, 4-benzoyl benzoic acid, pubchem14544, acmc-209mpx
PubChem CID 69147
IUPAC Name 4-benzoylbenzoic acid
SMILES C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B11645G View Documents TCI America
B11645G
5g Each for $125.28
 
B116425G View Documents TCI America
B116425G
25g Each for $380.42
 
Specifications

Chemical Identifiers

611-95-0
226.231
IFQUPKAISSPFTE-UHFFFAOYSA-N
69147
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C(=O)O
C14H10O3
MFCD00002560
p-benzoylbenzoic acid, benzoic acid, 4-benzoyl, 4-carboxybenzophenone, 4-benzoyl-benzoic acid, 4-phenylcarbonyl benzoic acid, benzoic acid, p-benzoyl, benzophenone-4-carboxylic acid, 4-benzoyl benzoic acid, pubchem14544, acmc-209mpx
4-benzoylbenzoic acid

Specifications

White
5g
C14H10O3
p-benzoylbenzoic acid, benzoic acid, 4-benzoyl, 4-carboxybenzophenone, 4-benzoyl-benzoic acid, 4-phenylcarbonyl benzoic acid, benzoic acid, p-benzoyl, benzophenone-4-carboxylic acid, 4-benzoyl benzoic acid, pubchem14544, acmc-209mpx
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C(=O)O
226.231
226.23
≥98.0% (T)
196°C
611-95-0
MFCD00002560
IFQUPKAISSPFTE-UHFFFAOYSA-N
4-benzoylbenzoic acid
69147
Crystal-Powder at 20°C
4-Benzoylbenzoic Acid
Safety and Handling

Safety and Handling

RTECSNumber : DG3650000

TSCA : No