Please login to your online account to display your discounted pricing

4-Benzoylbenzoic acid, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC105720250

 View more versions of this product

Catalog No. AC105720250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 226.23
Physical Form Crystalline Powder
Assay ≥95.5%
Quantity 25g
Color White to light beige
Melting Point 197° to 200°C
Beilstein 10, 753
CAS Max % 100.0
Linear Formula C6H5COC6H4CO2H
Molecular Formula C14H10O3
CAS 611-95-0
Chemical Name or Material 4-Benzoylbenzoic acid
Assay Percent Range 99%
MDL Number MFCD00002560
CAS Min % 98.5
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Benzoylbenzoic acid
CAS 611-95-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥95.5%
Assay Percent Range 99%
Molecular Formula C14H10O3
Linear Formula C6H5COC6H4CO2H
Formula Weight 226.23
MDL Number MFCD00002560
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents