Learn More
4-Benzoylbenzoic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1499314
Description
4-Benzoylbenzoic acid,is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Benzoylbenzoic acid,is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field etc.
Solubility
It is soluble in water (partly).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Incompatible with strong bases, strong oxidizing agents.
Specifications
4-Benzoylbenzoic acid | |
198°C to 200°C | |
C14H10O3 | |
25 g | |
p-benzoylbenzoic acid, benzoic acid, 4-benzoyl, 4-carboxybenzophenone, 4-benzoyl-benzoic acid, 4-phenylcarbonyl benzoic acid, benzoic acid, p-benzoyl, benzophenone-4-carboxylic acid, 4-benzoyl benzoic acid, pubchem14544, acmc-209mpx | |
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C(=O)O | |
226.231 | |
226.23 |
611-95-0 | |
Odorless | |
MFCD00002560 | |
1960224 | |
IFQUPKAISSPFTE-UHFFFAOYSA-N | |
4-benzoylbenzoic acid | |
69147 | |
99% |
Chemical Identifiers
611-95-0 | |
226.231 | |
IFQUPKAISSPFTE-UHFFFAOYSA-N | |
69147 | |
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C(=O)O |
C14H10O3 | |
MFCD00002560 | |
p-benzoylbenzoic acid, benzoic acid, 4-benzoyl, 4-carboxybenzophenone, 4-benzoyl-benzoic acid, 4-phenylcarbonyl benzoic acid, benzoic acid, p-benzoyl, benzophenone-4-carboxylic acid, 4-benzoyl benzoic acid, pubchem14544, acmc-209mpx | |
4-benzoylbenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Warning
RTECSNumber : DG3650000
RUO – Research Use Only