missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzoylbenzoic acid, 99%

Manufacturer:  Alfa Aesar™ A1499303

 View more versions of this product

Catalog No. AAA1499303



Description

Description

Applications
4-Benzoylbenzoic acid,is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field etc.

Solubility
It is soluble in water (partly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Incompatible with strong bases, strong oxidizing agents.
Specifications

Specifications

4-Benzoylbenzoic acid
198°C to 200°C
611-95-0
MFCD00002560
p-benzoylbenzoic acid, benzoic acid, 4-benzoyl, 4-carboxybenzophenone, 4-benzoyl-benzoic acid, 4-phenylcarbonyl benzoic acid, benzoic acid, p-benzoyl, benzophenone-4-carboxylic acid, 4-benzoyl benzoic acid, pubchem14544, acmc-209mpx
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C(=O)O
226.231
226.23
Odorless
1g
C14H10O3
1960224
IFQUPKAISSPFTE-UHFFFAOYSA-N
4-benzoylbenzoic acid
69147
99%

Chemical Identifiers

611-95-0
226.231
IFQUPKAISSPFTE-UHFFFAOYSA-N
69147
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C(=O)O
C14H10O3
MFCD00002560
p-benzoylbenzoic acid, benzoic acid, 4-benzoyl, 4-carboxybenzophenone, 4-benzoyl-benzoic acid, 4-phenylcarbonyl benzoic acid, benzoic acid, p-benzoyl, benzophenone-4-carboxylic acid, 4-benzoyl benzoic acid, pubchem14544, acmc-209mpx
4-benzoylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : DG3650000

Documents