Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzoylbenzoic acid, 99%

$31.65 - $322.45

Chemical Identifiers

Chemical Name or Material 4-Benzoylbenzoic acid
CAS 611-95-0
Molecular Formula C14H10O3
Formula Weight 226.23
MDL Number MFCD00002560
Beilstein 1960224
Synonym Benzophenone-4-carboxylic acid; 4-Carboxybenzophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1499303

  SDS
alfa aesar™
A1499303
1g Each for $31.65

AAA1499306

  SDS
alfa aesar™
A1499306
5g Each for $98.84

AAA1499314

  SDS
alfa aesar™
A1499314
25g Each for $322.45
Description & Specifications

Specifications

Formula Weight 226.23
Melting Point 198° to 200°C
Health Hazard 1 Warning
CAS 611-95-0
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Synonym Benzophenone-4-carboxylic acid; 4-Carboxybenzophenone
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number DG3650000
Odor Odorless
Molecular Formula C14H10O3
MDL Number MFCD00002560
Beilstein 1960224
Assay 99%
Chemical Name or Material 4-Benzoylbenzoic acid
Quantity 1g

Applications
4-Benzoylbenzoic acid,is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field etc.

Solubility
It is soluble in water (partly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Incompatible with strong bases, strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.