missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzoyl-3-methyl-1-phenyl-5-pyrazolone, TCI America™

$160.42 - $579.05

Chemical Identifiers

CAS 4551-69-3
Molecular Formula C17H14N2O2
MDL Number MFCD00003136
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B09765G View Documents TCI America
B09765G
5g Each for $160.42
 
B097625G View Documents TCI America
B097625G
25g Each for $579.05
 
Specifications

Chemical Identifiers

4551-69-3
C17H14N2O2
MFCD00003136

Specifications

White-Yellow
5g
C17H14N2O2
278.31
≥98.0% (HPLC,T)
118°C
4551-69-3
MFCD00003136
Crystal-Powder at 20°C
4-Benzoyl-3-methyl-1-phenyl-5-pyrazolone
Safety and Handling

Safety and Handling

TSCA : No