Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzoyl-3-methyl-1-phenyl-5-pyrazolinone, 98+%

$87.43 - $375.58

Chemical Identifiers

Chemical Name or Material 4-Benzoyl-3-methyl-1-phenyl-5-pyrazolinone
CAS 4551-69-3
Molecular Formula C17H14N2O2
Formula Weight 278.31
MDL Number MFCD00003136
Beilstein 888605
Synonym 4-Benzoyl-2,4-dihydro-5-methyl-2-phenyl-3H-pyrazol-3-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2253009

 
alfa aesar™
B2253009
10g Each for $87.43

AAB2253018

 
alfa aesar™
B2253018
50g Each for $375.58
Description & Specifications

Specifications

Formula Weight 278.31
Chemical Name or Material 4-Benzoyl-3-methyl-1-phenyl-5-pyrazolinone
Assay ≥98%
Molecular Formula C17H14N2O2
Melting Point 90° to 92°C
MDL Number MFCD00003136
Quantity 10g
CAS 4551-69-3
Beilstein 888605
Synonym 4-Benzoyl-2,4-dihydro-5-methyl-2-phenyl-3H-pyrazol-3-one