missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzamidopiperidine Hydrate, TCI America™

$172.64 - $514.88

Chemical Identifiers

CAS 33953-37-6
Molecular Formula C12H16N2O
Molecular Weight (g/mol) 204.273
MDL Number MFCD01314226
InChI Key JMQDNLCNCDSHNC-UHFFFAOYSA-N
Synonym n-piperidin-4-yl-benzamide, n-piperidin-4-yl benzamide, n-4-piperidyl benzamide, 4-benzamidopiperidine, n-4-piperidylbenzamide, benzamide, n-4-piperidinyl, 4-benzimido piperidine, n-4-piperidinyl benzamide, 4-benzamidopiperidine, 4-ben-zamidopiperidine
PubChem CID 118120
IUPAC Name N-piperidin-4-ylbenzamide
SMILES C1CNCCC1NC(=O)C2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B30255G View Documents TCI America
B30255G
5g Each for $172.64
 
B302525G View Documents TCI America
B302525G
25g Each for $514.88
 
Specifications

Chemical Identifiers

33953-37-6
204.273
JMQDNLCNCDSHNC-UHFFFAOYSA-N
118120
C1CNCCC1NC(=O)C2=CC=CC=C2
C12H16N2O
MFCD01314226
n-piperidin-4-yl-benzamide, n-piperidin-4-yl benzamide, n-4-piperidyl benzamide, 4-benzamidopiperidine, n-4-piperidylbenzamide, benzamide, n-4-piperidinyl, 4-benzimido piperidine, n-4-piperidinyl benzamide, 4-benzamidopiperidine, 4-ben-zamidopiperidine
N-piperidin-4-ylbenzamide

Specifications

White-Yellow
5g
C12H16N2O
n-piperidin-4-yl-benzamide, n-piperidin-4-yl benzamide, n-4-piperidyl benzamide, 4-benzamidopiperidine, n-4-piperidylbenzamide, benzamide, n-4-piperidinyl, 4-benzimido piperidine, n-4-piperidinyl benzamide, 4-benzamidopiperidine, 4-ben-zamidopiperidine
C1CNCCC1NC(=O)C2=CC=CC=C2
204.273
204.27
≥98.0% (GC,T)
137°C
33953-37-6
MFCD01314226
JMQDNLCNCDSHNC-UHFFFAOYSA-N
N-piperidin-4-ylbenzamide
118120
Crystal-Powder at 20°C
4-Benzamidopiperidine Hydrate
Safety and Handling

Safety and Handling

TSCA : No