Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzamidopiperidine hydrate, 98%

$66.15 - $186.84

Chemical Identifiers

Chemical Name or Material 4-Benzamidopiperidine hydrate
CAS 33953-37-6
Molecular Formula C12H16N2O·xH2O
Formula Weight 204.27 (Anhydrous)
MDL Number MFCD01314226
Synonym N-(4-Piperidinyl)benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2719303

  SDS
alfa aesar™
H2719303
1g Each for $66.15

AAH2719306

  SDS
alfa aesar™
H2719306
5g Each for $186.84
Description & Specifications

Specifications

Formula Weight 204.27 (Anhydrous)
Synonym N-(4-Piperidinyl)benzamide
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Chemical Name or Material 4-Benzamidopiperidine hydrate
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Assay 98%
CAS 33953-37-6
Melting Point 136° to 146°C
Molecular Formula C12H16N2O·xH2O
MDL Number MFCD01314226
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.