missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Azepanone hydrochloride, 96%

$65.82 - $1,009.61

Chemical Identifiers

CAS 50492-22-3
Molecular Formula C6H12ClNO
Molecular Weight (g/mol) 149.618
MDL Number MFCD02183487
InChI Key WFTRLIZPJMFJER-UHFFFAOYSA-N
Synonym 4-azepanone hydrochloride, 4-oxohomopiperidine-hcl, 4-perhydroazepinone hcl, 4-perhydroazepinone hydrochloride, 4h-azepin-4-one, hexahydro-, hydrochloride, azepan-4-one hydrochloride, hexahydro-4-azepinone hydrochloride, hexahydro-4h-azepin-4-one hcl, hexahydro-4h-azepin-4-one hydrochloride, pubchem17662
PubChem CID 22021728
IUPAC Name azepan-4-one;hydrochloride
SMILES C1CC(=O)CCNC1.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6635503
View Documents
Alfa Aesar™
H6635503
1g Each for $65.82
 
AAH6635506
View Documents
Alfa Aesar™
H6635506
5g Each for $246.24
 
AAH6635514
View Documents
Alfa Aesar™
H6635514
25g Each for $1,009.61
 
Specifications

Chemical Identifiers

50492-22-3
149.618
WFTRLIZPJMFJER-UHFFFAOYSA-N
22021728
C1CC(=O)CCNC1.Cl
C6H12ClNO
MFCD02183487
4-azepanone hydrochloride, 4-oxohomopiperidine-hcl, 4-perhydroazepinone hcl, 4-perhydroazepinone hydrochloride, 4h-azepin-4-one, hexahydro-, hydrochloride, azepan-4-one hydrochloride, hexahydro-4-azepinone hydrochloride, hexahydro-4h-azepin-4-one hcl, hexahydro-4h-azepin-4-one hydrochloride, pubchem17662
azepan-4-one;hydrochloride

Specifications

152°C (306°F)
1g
C6H12ClNO
3687441
WFTRLIZPJMFJER-UHFFFAOYSA-N
azepan-4-one;hydrochloride
22021728
Solid
4-Azepanone hydrochloride
176°C to 181°C
50492-22-3
MFCD02183487
4-azepanone hydrochloride, 4-oxohomopiperidine-hcl, 4-perhydroazepinone hcl, 4-perhydroazepinone hydrochloride, 4h-azepin-4-one, hexahydro-, hydrochloride, azepan-4-one hydrochloride, hexahydro-4-azepinone hydrochloride, hexahydro-4h-azepin-4-one hcl, hexahydro-4h-azepin-4-one hydrochloride, pubchem17662
C1CC(=O)CCNC1.Cl
149.618
149.62
96%
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302-H315-H318-H335

TSCA : No

Recommended Storage : Ambient temperatures