Learn More
4-Azaindole, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6422403
Description
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in methanol and chloroform. Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
4-Azaindole | |
125°C to 127°C | |
MFCD00971977 | |
109683 | |
Soluble in methanol and chloroform. Slightly soluble in water. | |
N1C=CC2=NC=CC=C12 | |
118.14 | |
118.14 | |
Solid |
272-49-1 | |
C7H6N2 | |
1 g | |
4-azaindole, 1h-pyrrolo 3,2-b pyridine, 1,4-diazaindene, pyrrolopyridine, 4h-pyrrolo 3,2-b pyridine, 1h-pyrrolo 3.2-b pyridine, zlchem 17, pubchem7675, acmc-1clkx | |
XWIYUCRMWCHYJR-UHFFFAOYSA-N | |
1H-pyrrolo[3,2-b]pyridine | |
9226 | |
97% |
Chemical Identifiers
272-49-1 | |
118.14 | |
XWIYUCRMWCHYJR-UHFFFAOYSA-N | |
9226 | |
N1C=CC2=NC=CC=C12 |
C7H6N2 | |
MFCD00971977 | |
4-azaindole, 1h-pyrrolo 3,2-b pyridine, 1,4-diazaindene, pyrrolopyridine, 4h-pyrrolo 3,2-b pyridine, 1h-pyrrolo 3.2-b pyridine, zlchem 17, pubchem7675, acmc-1clkx | |
1H-pyrrolo[3,2-b]pyridine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : UY8715000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only