Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Anilino-1-methylpiperidine, 98%

$55.55 - $464.86

Chemical Identifiers

Chemical Name or Material 4-Anilino-1-methylpiperidine
CAS 22261-94-5
Molecular Formula C12H18N2
Formula Weight 190.28
MDL Number MFCD06408133
Synonym 1-Benzyl-N-methyl-4-piperidinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2727603

  SDS
alfa aesar™
H2727603
1g Each for $55.55

AAH2727606

  SDS
alfa aesar™
H2727606
5g Each for $153.93

AAH2727614

  SDS
alfa aesar™
H2727614
25g Each for $464.86
Description & Specifications

Specifications

Formula Weight 190.28
Chemical Name or Material 4-Anilino-1-methylpiperidine
Assay 98%
Molecular Formula C12H18N2
MDL Number MFCD06408133
CAS 22261-94-5
Synonym 1-Benzyl-N-methyl-4-piperidinamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 82° to 86°C
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.