Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Anilino-1-benzylpiperidine, 98%

$54.39 - $455.04

Chemical Identifiers

Chemical Name or Material 4-Anilino-1-benzylpiperidine
CAS 1155-56-2
Molecular Formula C18H22N2
Formula Weight 266.38
MDL Number MFCD00023738
Synonym 1-Benzyl-N-phenyl-4-piperidinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2715803

  SDS
alfa aesar™
H2715803
1g Each for $54.39

AAH2715814

  SDS
alfa aesar™
H2715814
25g Each for $455.04

AAH2715806

  SDS
alfa aesar™
H2715806
5g Each for $150.68
Description & Specifications

Specifications

Formula Weight 266.38
Chemical Name or Material 4-Anilino-1-benzylpiperidine
Assay 98%
Molecular Formula C18H22N2
MDL Number MFCD00023738
CAS 1155-56-2
Synonym 1-Benzyl-N-phenyl-4-piperidinamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
4-Anilino-1-benzylpiperidine, is used as a pharmaceutical intermediate.

Solubility
Soluble in dichloromethane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.