Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Aminothiobenzamide, 97%

$232.61 - $232.61

Chemical Identifiers

Chemical Name or Material 4-Aminothiobenzamide
CAS 4114-67-4
Molecular Formula C7H8N2S
Formula Weight 152.21g/mol
MDL Number MFCD00040927
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5184906

 
alfa aesar™
H51849-06
5g Each for $232.61
Description & Specifications

Specifications

Quantity 5g
Melting Point 185° to 190°C
Quantity 5g
Chemical Name or Material 4-Aminothiobenzamide
CAS 4114-67-4
Assay 97%
Molecular Formula C7H8N2S
Formula Weight 152.21g/mol
MDL Number MFCD00040927
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.