Please login to your online account to display your discounted pricing

4-Aminotetrahydropyran, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC29914CB

 View more versions of this product

Catalog No. CC29914CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 101.15
Quantity 250mg
Assay 97%
CAS Max % 100.0
Chemical Name or Material 4-Aminotetrahydropyran
MDL Number MFCD02179436
CAS 38041-19-9
CAS Min % 96.5
Assay Percent Range 97%
Molecular Formula C5H11NO
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Causes skin irritation. May cause respiratory irritation. Harmful if swallowed. Causes serious eye irritation.
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remov
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 4-Aminotetrahydropyran
CAS 38041-19-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H11NO
Formula Weight 101.15
MDL Number MFCD02179436
Accessories

Danger

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remov

GHS P Statement Causes skin irritation. May cause respiratory irritation. Harmful if swallowed. Causes serious eye irritation.