Please login to your online account to display your discounted pricing

4-Aminotetrahydropyran, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-Aminotetrahydropyran
CAS 38041-19-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H11NO
Formula Weight 101.15
MDL Number MFCD02179436
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC29914CB

 
fisher scientific company
CC29914CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC29914DA

 
fisher scientific company
CC29914DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 101.15
Quantity 250mg
Assay 97%
CAS Max % 100.0
Chemical Name or Material 4-Aminotetrahydropyran
MDL Number MFCD02179436
CAS 38041-19-9
CAS Min % 96.5
Assay Percent Range 97%
Molecular Formula C5H11NO
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Causes skin irritation. May cause respiratory irritation. Harmful if swallowed. Causes serious eye irritation.
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remov
Packaging Amber Glass Bottle
Safety and Handling

Danger

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remov

GHS P Statement Causes skin irritation. May cause respiratory irritation. Harmful if swallowed. Causes serious eye irritation.