Learn More
4-Aminopyridine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 104570050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Aminopyridine | |
504-24-5 | |
100.0 | |
White to Yellow | |
156°C | |
97.5% min. (GC) | |
C5H6N2 | |
MFCD00006439 | |
22,433 | |
4-aminopyridine, fampridine, 4-pyridinamine, dalfampridine, 4-pyridylamine, avitrol, p-aminopyridine, 4-ap, ampyra, gamma-aminopyridine | |
NUKYPUAOHBNCPY-UHFFFAOYSA-N | |
pyridin-4-amine | |
1727 | |
94.12 | |
Crystalline Powder, Crystals or Solid |
98% | |
97.5 | |
157°C to 161°C | |
273°C | |
Authentic | |
Glass bottle | |
(C5H4N)NH2 | |
5 g | |
15,2802 | |
Solubility in water: 74g/L water (20°C). Other solubilities: soluble in ethanol,benzene and ether,slightly soluble in acetone and methanol | |
C1=CN=CC=C1N | |
94.12 | |
CHEBI:34385 | |
98% |
Chemical Identifiers
504-24-5 | |
94.12 | |
NUKYPUAOHBNCPY-UHFFFAOYSA-N | |
1727 | |
pyridin-4-amine |
C5H6N2 | |
MFCD00006439 | |
4-aminopyridine, fampridine, 4-pyridinamine, dalfampridine, 4-pyridylamine, avitrol, p-aminopyridine, 4-ap, ampyra, gamma-aminopyridine | |
CHEBI:34385 | |
C1=CN=CC=C1N |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Fatal if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 207-987-9
RTECSNumber : US1750000
TSCA : TSCA
RUO – Research Use Only