Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Aminopyridine-3-sulfonic acid monohydrate, 98%

$51.88 - $424.84

Chemical Identifiers

Chemical Name or Material 4-Aminopyridine-3-sulfonic acid monohydrate
CAS 29452-57-1
Molecular Formula C5H6N2O3S·H2O
Formula Weight 192.20 (174.17 Anhydrous)
MDL Number MFCD09953683
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2595503

  SDS
alfa aesar™
H2595503
1g Each for $51.88

AAH2595506

  SDS
alfa aesar™
H2595506
5g Each for $142.07

AAH2595514

  SDS
alfa aesar™
H2595514
25g Each for $424.84
Description & Specifications

Specifications

Formula Weight 192.20 (174.17 Anhydrous)
Assay 98%
MDL Number MFCD09953683
Chemical Name or Material 4-Aminopyridine-3-sulfonic acid monohydrate
Melting Point ≈320°C
Quantity 1g
Molecular Formula C5H6N2O3S·H2O
CAS 29452-57-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.