Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Aminopyridine-3-carboxaldehyde, tech. 85%

$244.74 - $1,183.70

Chemical Identifiers

Chemical Name or Material 4-Aminopyridine-3-carboxaldehyde
CAS 42373-30-8
Purity Grade Technical Grade
Molecular Formula C6H6N2O
Formula Weight 122.13
MDL Number MFCD04038972
Synonym 4-Aminonicotinaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5002803

 
alfa aesar™
H5002803
1g Each for $244.74

AAH5002806

 
alfa aesar™
H5002806
5g Each for $1,183.70
Description & Specifications

Specifications

Formula Weight 122.13
Chemical Name or Material 4-Aminopyridine-3-carboxaldehyde
Melting Point 108° to 112°C
Purity Grade Technical Grade
Quantity 1g
Assay 85%
Molecular Formula C6H6N2O
MDL Number MFCD04038972
CAS 42373-30-8
Synonym 4-Aminonicotinaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.