missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Aminopyridine-3-carboxaldehyde, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC437350050
Specifications
4-Aminopyridine-3-carboxaldehyde | |
Cream to White | |
5g | |
94.0 | |
94% min. (HPLC) | |
3-pyridinecarboxaldehyde, 4-amino, 4-amino-3-formylpyridine, 4-amino-3-pyridinecarboxaldehyde, 4-amino-pyridine-3-carbaldehyde, 4-amino-pyridine-3-carboxaldehyde, 4-aminonicotinaldehyde, 4-aminopyridine-3-carboxaldehyde, acmc-1aqvh, pubchem7681, zlchem 837 | |
C1=CN=CC(=C1N)C=O | |
122.13 | |
122.13 | |
95% |
Authentic | |
111.2°C to 112.7°C | |
42373-30-8 | |
100.0 | |
C6H6N2O | |
GTPZHMGXKZIHKW-UHFFFAOYSA-N | |
4-aminopyridine-3-carbaldehyde | |
2761033 | |
Powder |
Chemical Identifiers
42373-30-8 | |
122.13 | |
3-pyridinecarboxaldehyde, 4-amino, 4-amino-3-formylpyridine, 4-amino-3-pyridinecarboxaldehyde, 4-amino-pyridine-3-carbaldehyde, 4-amino-pyridine-3-carboxaldehyde, 4-aminonicotinaldehyde, 4-aminopyridine-3-carboxaldehyde, acmc-1aqvh, pubchem7681, zlchem 837 | |
4-aminopyridine-3-carbaldehyde |
C6H6N2O | |
GTPZHMGXKZIHKW-UHFFFAOYSA-N | |
2761033 | |
C1=CN=CC(=C1N)C=O |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning