Please login to your online account to display your discounted pricing

4'-Aminopropiophenone, 98%, ACROS Organics™


Quantity 50g

Manufacturer: thermo fisher scientific  AC401040500

 View more versions of this product

Catalog No. AC401040500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 149.19
Quantity 50g
Chemical Name or Material 4'-Aminopropiophenone
CAS 70-69-9
Molecular Formula C9H11NO
MDL Number MFCD00017113
Assay Percent Range 98%
Synonym 1-(4-Aminophenyl)-1-propanone

Specifications

Chemical Name or Material 4'-Aminopropiophenone
Assay Percent Range 98%
Molecular Formula C9H11NO
Formula Weight 149.19
MDL Number MFCD00017113
Synonym 1-(4-Aminophenyl)-1-propanone