missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Aminophthalonitrile, 98%

Manufacturer:  Alfa Aesar™ L1343306

 View more versions of this product

Catalog No. AAL1343306



Specifications

Specifications

4-Aminophthalonitrile
5g
C8H5N3
UN3439
4-aminophthalonitrile, 3,4-dicyanoaniline, 1,2-benzenedicarbonitrile, 4-amino, 3,4-dicyanoanline, 4-amino-1,2-benzenedicarbonitrile, 4-amino-phthalonitril, 4-amino-phthalonitrile, 3,4-dicyano aniline, acmc-1awu3, 4-aminophthalonitrile
C1=CC(=C(C=C1N)C#N)C#N
143.149
143.15
179°C to 182°C
56765-79-8
MFCD00041741
2937382
RRCAJFYQXKPXOJ-UHFFFAOYSA-N
4-aminobenzene-1,2-dicarbonitrile
92529
98%

Chemical Identifiers

56765-79-8
143.149
RRCAJFYQXKPXOJ-UHFFFAOYSA-N
92529
C1=CC(=C(C=C1N)C#N)C#N
C8H5N3
MFCD00041741
4-aminophthalonitrile, 3,4-dicyanoaniline, 1,2-benzenedicarbonitrile, 4-amino, 3,4-dicyanoanline, 4-amino-1,2-benzenedicarbonitrile, 4-amino-phthalonitril, 4-amino-phthalonitrile, 3,4-dicyano aniline, acmc-1awu3, 4-aminophthalonitrile
4-aminobenzene-1,2-dicarbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.

GHS P Statement
P280-P301+P310-P361-P312-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : TSCA

Documents