Learn More
4-Aminophthalonitrile, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 401020250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Aminophthalonitrile | |
56765-79-8 | |
100.0 | |
Orange | |
96% min. (HPLC) | |
C8H5N3 | |
25 g | |
4-aminophthalonitrile, 3,4-dicyanoaniline, 1,2-benzenedicarbonitrile, 4-amino, 3,4-dicyanoanline, 4-amino-1,2-benzenedicarbonitrile, 4-amino-phthalonitril, 4-amino-phthalonitrile, 3,4-dicyano aniline, acmc-1awu3 | |
RRCAJFYQXKPXOJ-UHFFFAOYSA-N | |
4-aminobenzene-1,2-dicarbonitrile | |
92529 | |
97% |
97% | |
96.0 | |
177.0°C to 182.0°C | |
Authentic | |
Glass bottle | |
MFCD00041741 | |
05, | |
Solubility in water: insoluble. Other solubilities: soluble in methanol | |
C1=CC(=C(C=C1N)C#N)C#N | |
143.15 | |
143.15 | |
Crystalline Powder |
Chemical Identifiers
56765-79-8 | |
143.15 | |
RRCAJFYQXKPXOJ-UHFFFAOYSA-N | |
92529 | |
C1=CC(=C(C=C1N)C#N)C#N |
C8H5N3 | |
MFCD00041741 | |
4-aminophthalonitrile, 3,4-dicyanoaniline, 1,2-benzenedicarbonitrile, 4-amino, 3,4-dicyanoanline, 4-amino-1,2-benzenedicarbonitrile, 4-amino-phthalonitril, 4-amino-phthalonitrile, 3,4-dicyano aniline, acmc-1awu3 | |
4-aminobenzene-1,2-dicarbonitrile |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a po
GHS Signal Word: Danger
EINECSNumber : 260-370-6
RUO – Research Use Only