missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Aminophthalimide 97%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 401010010

 View more versions of this product

Catalog No. AC401010010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
Powder
Yellow-brown
100.0
Authentic
97%
3676-85-5
97%
162.15
1g
96.0
22, V,13, 268
4-Aminophthalimide
MFCD00041854
C8H6N2O2
Glass bottle

Chemical Identifiers

3676-85-5
100.0
C8H6N2O2
MFCD00041854
≥96.0%
96.0
97%
162.15
4-Aminophthalimide, 97%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 222-948-6

TSCA : TSCA