Please login to your online account to display your discounted pricing

4-Aminophthalimide 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Aminophthalimide
Name Note 97%
CAS 3676-85-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H6N2O2
Formula Weight 162.15
MDL Number MFCD00041854
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC401010010

  SDS
fisher scientific acros
401010010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 162.15
Physical Form Powder
Quantity 1g
Color Yellow-brown
TSCA TSCA
EINECS Number 222-948-6
CAS Min % 96.0
CAS Max % 100.0
Beilstein 22, V,13, 268
Infrared Spectrum Authentic
Chemical Name or Material 4-Aminophthalimide
Assay Percent Range 97%
MDL Number MFCD00041854
CAS 3676-85-5
Molecular Formula C8H6N2O2
Name Note 97%
Packaging Glass bottle