Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Aminophenylthio)acetic acid, 97%

$20.04 - $86.19

Chemical Identifiers

Chemical Name or Material (4-Aminophenylthio)acetic acid
CAS 104-18-7
Molecular Formula C8H9NO2S
Formula Weight 183.23
MDL Number MFCD00025360
Beilstein 2720454
Synonym 4-Aminothiophenoxyacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1983403

  SDS
alfa aesar™
A1983403
1g Each for $20.04

AAA1983409

  SDS
alfa aesar™
A1983409
10g Each for $86.19
Description & Specifications

Specifications

Formula Weight 183.23
CAS 104-18-7
Synonym 4-Aminothiophenoxyacetic acid
Assay 97%
Quantity 1g
Beilstein 2720454
Melting Point ≈200°C (dec.)
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00025360
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Chemical Name or Material (4-Aminophenylthio)acetic acid
Molecular Formula C8H9NO2S

Applications
It can react with 2-phenyl-benzo[d][1,3]oxazin-4-one to get 2-(Phenyl-4-(3H)-quinazolyl)-phenyl-4′-mercaptoacetic acid. This reaction needs solvents pyridine and H2O by heating.

Solubility
Soluble in methanol, acetone, toluene, n-octanol, acetonitrile, ethyl acetate, dichloromethane.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.