Please login to your online account to display your discounted pricing

4-Aminophenylacetic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Aminophenylacetic acid
CAS 1197-55-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (on anhydrous substance)
Assay Percent Range 98%
Molecular Formula C8H9NO2
Linear Formula H2NC6H4CH2CO2H
Formula Weight 151.16
MDL Number MFCD00007916
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC104281000

  SDS
thermo fisher scientific
AC104281000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC104280050

  SDS
fisher scientific acros
104280050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC104280250

  SDS
fisher scientific acros
104280250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 151.16
Physical Form Powder
Assay ≥97.5% (on anhydrous substance)
Quantity 100g
Color Light yellow to beige
Melting Point 196° to 201°C
Beilstein 14, 456
Merck Index 15, 459
CAS Max % 100.0
Water 1% max. (K.F.)
Individual Impurity 4-nitrophenylacetic acid (HPLC): < 0.5%
Linear Formula H2NC6H4CH2CO2H
CAS 1197-55-3
Assay Percent Range 98%
Chemical Name or Material 4-Aminophenylacetic acid
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00007916
CAS Min % 97.5
Packaging Glass bottle
Molecular Formula C8H9NO2
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.