Learn More
4-Aminophenylacetic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 104280050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Aminophenylacetic acid | |
97.5 | |
97.5% min. (HPLC) | |
Authentic | |
MFCD00007916 | |
15, 459 | |
4-aminophenylacetic acid, 2-4-aminophenyl acetic acid, benzeneacetic acid, 4-amino, p-aminophenylacetic acid, 4-aminobenzeneacetic acid, 4-carboxymethylaniline, acetic acid, p-aminophenyl, para-aminophenyl acetic acid, p-aminophenyl acetic acid, 4-aminophenyl acetic acid | |
CSEWAUGPAQPMDC-UHFFFAOYSA-N | |
2-(4-aminophenyl)acetic acid | |
14533 | |
98% | |
5g |
1197-55-3 | |
100.0 | |
C8H9NO2 | |
H2NC6H4CH2CO2H | |
14, 456 | |
Glass bottle | |
Solubility in water: insoluble. Other solubilities: soluble in alkalies, soluble in alcohol | |
NC1=CC=C(CC(O)=O)C=C1 | |
151.17 | |
151.16 | |
1% max. (K.F.) |
Chemical Identifiers
1197-55-3 | |
151.17 | |
CSEWAUGPAQPMDC-UHFFFAOYSA-N | |
14533 | |
NC1=CC=C(CC(O)=O)C=C1 |
C8H9NO2 | |
MFCD00007916 | |
4-aminophenylacetic acid, 2-4-aminophenyl acetic acid, benzeneacetic acid, 4-amino, p-aminophenylacetic acid, 4-aminobenzeneacetic acid, 4-carboxymethylaniline, acetic acid, p-aminophenyl, para-aminophenyl acetic acid, p-aminophenyl acetic acid, 4-aminophenyl acetic acid | |
2-(4-aminophenyl)acetic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 214-828-7
RUO â Research Use Only