Learn More
4-Aminophenylacetic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1467509
Description
4-Aminophenylacetic acid, is used as a non-translocated competitive inhibitor of the epithelial peptide transporter PepT1. It can be detected using HPLC, NMR techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Aminophenylacetic acid, is used as a non-translocated competitive inhibitor of the epithelial peptide transporter PepT1. It can be detected using HPLC, NMR techniques.
Solubility
Very soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents.Stable under normal temperatures and pressures. Stable at room temperature in closed containers under normal storage and handling conditions.
Specifications
∼201°C (decomposition) | |
1197-55-3 | |
MFCD00007916 | |
14,463 | |
Very soluble in water. | |
NC1=CC=C(CC(O)=O)C=C1 | |
151.17 | |
151.17 | |
10 g |
4-Aminophenylacetic acid | |
C8H9NO2 | |
2208094 | |
4-aminophenylacetic acid, 2-4-aminophenyl acetic acid, benzeneacetic acid, 4-amino, p-aminophenylacetic acid, 4-aminobenzeneacetic acid, 4-carboxymethylaniline, acetic acid, p-aminophenyl, para-aminophenyl acetic acid, p-aminophenyl acetic acid, 4-aminophenyl acetic acid | |
CSEWAUGPAQPMDC-UHFFFAOYSA-N | |
2-(4-aminophenyl)acetic acid | |
14533 | |
98% |
Chemical Identifiers
1197-55-3 | |
151.17 | |
CSEWAUGPAQPMDC-UHFFFAOYSA-N | |
14533 | |
NC1=CC=C(CC(O)=O)C=C1 |
C8H9NO2 | |
MFCD00007916 | |
4-aminophenylacetic acid, 2-4-aminophenyl acetic acid, benzeneacetic acid, 4-amino, p-aminophenylacetic acid, 4-aminobenzeneacetic acid, 4-carboxymethylaniline, acetic acid, p-aminophenyl, para-aminophenyl acetic acid, p-aminophenyl acetic acid, 4-aminophenyl acetic acid | |
2-(4-aminophenyl)acetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-828-7
RTECSNumber : AF3550000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only