Please login to your online account to display your discounted pricing

4-Aminophenyl Sulfone 97%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  104360050

 View more versions of this product

Catalog No. AC104360050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 248.3
Quantity 5g
Color White to beige
EINECS Number 201-248-4
TSCA TSCA
Melting Point 173° to 178°C
RTECS Number BY8925000
Beilstein 13, 536
Merck Index 15, 2822
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-Aminophenyl sulfone
Name Note 97%
Assay Percent Range 97%
Molecular Formula C12H12N2O2S
Linear Formula (H2NC6H4)2SO2
MDL Number MFCD00007887
CAS 80-08-0
Infrared Spectrum Authentic
Synonym 4, 4'-Sulfonyldianiline
Packaging Glass bottle
Health Hazard 1 CANCER SUSPECTED AGENT
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Aminophenyl sulfone
Name Note 97%
CAS 80-08-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C12H12N2O2S
Linear Formula (H2NC6H4)2SO2
Formula Weight 248.3
MDL Number MFCD00007887
Synonym 4, 4'-Sulfonyldianiline
Accessories

CANCER SUSPECTED AGENT

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents