Learn More
4-Aminophenyl sulfone, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 104361000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Aminophenyl sulfone | |
80-08-0 | |
100.0 | |
97% | |
C12H12N2O2S | |
MFCD00007887 | |
13, 536 | |
dapsone, diaphenylsulfone, 4,4'-diaminodiphenyl sulfone, dapson, 4,4'-sulfonyldianiline, diaphenylsulfon, sulfona, 4-aminophenyl sulfone, avlosulfone, diphenasone | |
MQJKPEGWNLWLTK-UHFFFAOYSA-N | |
4-(4-aminophenyl)sulfonylaniline | |
2955 | |
248.3 |
97% | |
96.0 | |
Authentic | |
Glass bottle | |
(H2NC6H4)2SO2 | |
100g | |
15, 2822 | |
Solubility in water: insoluble. Other solubilities: soluble in diluted hydrochloric acid,soluble in alcohol,methanol and acetone | |
C1=CC(=CC=C1N)S(=O)(=O)C2=CC=C(C=C2)N | |
248.3 | |
CHEBI:4325 | |
97% |
Chemical Identifiers
80-08-0 | |
248.3 | |
MQJKPEGWNLWLTK-UHFFFAOYSA-N | |
2955 | |
4-(4-aminophenyl)sulfonylaniline |
C12H12N2O2S | |
MFCD00007887 | |
dapsone, diaphenylsulfone, 4,4'-diaminodiphenyl sulfone, dapson, 4,4'-sulfonyldianiline, diaphenylsulfon, sulfona, 4-aminophenyl sulfone, avlosulfone, diphenasone | |
CHEBI:4325 | |
C1=CC(=CC=C1N)S(=O)(=O)C2=CC=C(C=C2)N |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 201-248-4
RTECSNumber : BY8925000
TSCA : TSCA
RUO â Research Use Only