Please login to your online account to display your discounted pricing

4-Aminophenyl ether, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC104331000

 View more versions of this product

Catalog No. AC104331000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 200.24
Quantity 100g
Beilstein 13, 441
CAS Max % 100.0
Linear Formula (H2NC6H4)2O
Synonym 4, 4';-Diaminodiphenyl ether
Molecular Formula C12H12N2O
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause cancer.
May cause genetic defects.
Suspected of damaging fertility.
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.
CAS Min % 97.5
Packaging Glass bottle
Chemical Name or Material 4-Aminophenyl ether
MDL Number MFCD00007863
Assay Percent Range 98%
CAS 101-80-4

Specifications

Chemical Name or Material 4-Aminophenyl ether
CAS 101-80-4
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C12H12N2O
Linear Formula (H2NC6H4)2O
Formula Weight 200.24
MDL Number MFCD00007863
Synonym 4, 4';-Diaminodiphenyl ether
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause cancer.
May cause genetic defects.
Suspected of damaging fertility.
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.

SDS & Safety Documents