missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Aminophenol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 104272500

 View more versions of this product

Catalog No. AC104272500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Aminophenol
96.0
187.0°C to 191.0°C
284.0°C
Authentic
Glass bottle
H2NC6H4OH
13,427
p-aminophenol, 4-hydroxyaniline, p-hydroxyaniline, phenol, 4-amino, paranol, 4-aminobenzenol, certinal, citol, azol, fouramine p
PLIKAWJENQZMHA-UHFFFAOYSA-N
4-aminophenol
403
109.13
Crystalline Powder
123-30-8
100.0
White to Cream
189°C
96% min. (GC)
C6H7NO
250 g
15,458
Solubility in water: 15g/L (20°C). Other solubilities: soluble in alcohol, practically insoluble in benzene and chloroform, 9.3% in ethyl methyl ketone (58.5°C), 4.5% in abs.ethanol (0°C)
C1=CC(=CC=C1N)O
109.13
CHEBI:17602
97%

Chemical Identifiers

123-30-8
109.13
p-aminophenol, 4-hydroxyaniline, p-hydroxyaniline, phenol, 4-amino, paranol, 4-aminobenzenol, certinal, citol, azol, fouramine p
CHEBI:17602
C1=CC(=CC=C1N)O
C6H7NO
PLIKAWJENQZMHA-UHFFFAOYSA-N
403
4-aminophenol
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 204-616-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only