Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Aminomethyl)pyridine, 97+%

Chemical Identifiers

Chemical Name or Material 4-(Aminomethyl)pyridine
CAS 3731-53-1
Molecular Formula C6H8N2
Formula Weight 108.14
MDL Number MFCD00006449
UN Number UN2735
Beilstein 108057
Synonym 4-Picolylamine; 4-Pyridinemethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0634514

 
alfa aesar™
L0634514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0634522

 
alfa aesar™
L0634522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 108.14
Chemical Name or Material 4-(Aminomethyl)pyridine
TSCA TSCA
Assay ≥97%
Molecular Formula C6H8N2
MDL Number MFCD00006449
Quantity 25g
Beilstein 108057
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Melting Point −8°C
Boiling Point 230°C
CAS 3731-53-1
UN Number UN2735
Flash Point 108°C (226°F)
Odor Amine-like
Synonym 4-Picolylamine; 4-Pyridinemethylamine
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.