Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Aminomethyl)piperidine, 98+%

Chemical Identifiers

Chemical Name or Material 4-(Aminomethyl)piperidine
CAS 7144-05-0
Molecular Formula C6H14N2
Formula Weight 114.19
MDL Number MFCD00006007
UN Number UN3259
Beilstein 471185
Synonym 4-Piperidinemethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1157706

 
alfa aesar™
L1157706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1157714

 
alfa aesar™
L1157714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 114.19
Chemical Name or Material 4-(Aminomethyl)piperidine
MDL Number MFCD00006007
CAS 7144-05-0
TSCA TSCA
Assay ≥98%
Molecular Formula C6H14N2
Boiling Point 199° to 200°C
Beilstein 471185
Flash Point 79°C (174°F)
Synonym 4-Piperidinemethylamine
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
Melting Point 23° to 25°C
UN Number UN3259
Quantity 5g
Health Hazard 1 Danger

Applications
4-(Aminomethyl)piperidine is used as a reagent for rapid cleavage of the Fmoc group in peptide synthesis, particularly recommended in conjunction with Fmoc-protected acid chlorides as the active species. It Is also used as a reactant for evaulated as a small molecule inhibitor of CD4-gp120 binding and as a reagent involved in solvothermal preparation of germanium oxide hydroxide 3D net zeotype, involved in zeolite synthesis and as a coupling agent for adhesive systems.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.