missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Aminomethyl)indole, 97%

Manufacturer:  Alfa Aesar™ H54963MD

 View more versions of this product

Catalog No. AAH54963MD



Description

Description

Applications
4-(Aminomethyl)indole is used as a reactant for preparation of dopamine receptor antagonists and for preparation of high-affinity ligands to α2δ subunit of voltage-gated calcium channels. It is used as a synthetic intermediate.

Solubility
Soluble in ethanol, dimethyl sulfoxide and dimethyl formamide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

4-(Aminomethyl)indole
250mg
C9H10N2
1-1h-indol-4-yl methanamine, 1h-indol-4-yl methanamine, 1h-indol-4-ylmethyl amine, 1h-indole-4-methanamine, 4-aminomethyl indole, 4-aminomethyl-1h-indole, 4-aminomethyl-indole, 4-aminomethylindole, c-1h-indol-4-yl-methylamine, indole-4-methylamine
FFBWKPKOXRMLNP-UHFFFAOYSA-N
1H-indol-4-ylmethanamine
280302
97%
130°C to 135°C
3468-18-6
MFCD04973297
Soluble in ethanol,dimethyl sulfoxide and dimethyl formamide.
NCC1=C2C=CNC2=CC=C1
146.19
146.19

Chemical Identifiers

3468-18-6
146.19
FFBWKPKOXRMLNP-UHFFFAOYSA-N
280302
NCC1=C2C=CNC2=CC=C1
C9H10N2
MFCD04973297
1-1h-indol-4-yl methanamine, 1h-indol-4-yl methanamine, 1h-indol-4-ylmethyl amine, 1h-indole-4-methanamine, 4-aminomethyl indole, 4-aminomethyl-1h-indole, 4-aminomethyl-indole, 4-aminomethylindole, c-1h-indol-4-yl-methylamine, indole-4-methylamine
1H-indol-4-ylmethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents
Product Certifications