missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Aminomethyl)indole, 97%

Manufacturer:  Alfa Aesar™ H5496306

 View more versions of this product

Catalog No. AAH5496306



Description

Description

Applications
4-(Aminomethyl)indole is used as a reactant for preparation of dopamine receptor antagonists and for preparation of high-affinity ligands to α2δ subunit of voltage-gated calcium channels. It is used as a synthetic intermediate.

Solubility
Soluble in ethanol, dimethyl sulfoxide and dimethyl formamide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

4-(Aminomethyl)indole
5g
C9H10N2
1-1h-indol-4-yl methanamine
FFBWKPKOXRMLNP-UHFFFAOYSA-N
1H-indol-4-ylmethanamine
280302
97%
130°C to 135°C
3468-18-6
MFCD04973297
Soluble in ethanol,dimethyl sulfoxide and dimethyl formamide.
C1=CC(=C2C=CNC2=C1)CN
146.193
146.19

Chemical Identifiers

3468-18-6
146.193
FFBWKPKOXRMLNP-UHFFFAOYSA-N
280302
C1=CC(=C2C=CNC2=C1)CN
C9H10N2
MFCD04973297
1-1h-indol-4-yl methanamine
1H-indol-4-ylmethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents