Please login to your online account to display your discounted pricing

4-Aminomethyl-2-chloropyridine, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Aminomethyl-2-chloropyridine
CAS 144900-57-2
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94.0%
Assay Percent Range 95%
Molecular Formula C6H7ClN2
Formula Weight 142.59
Synonym 2-Chloro-4-(aminomethyl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC438890050

 
thermo fisher scientific
AC438890050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC438890010

 
thermo fisher scientific
AC438890010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 142.59
Physical Form Liquid
Assay ≥94.0%
Quantity 5g
Color Colorless to yellow
CAS Max % 100.0
CAS Min % 94.0
Chemical Name or Material 4-Aminomethyl-2-chloropyridine
Molecular Formula C6H7ClN2
Assay Percent Range 95%
CAS 144900-57-2
Infrared Spectrum Authentic
Synonym 2-Chloro-4-(aminomethyl)pyridine
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.