Learn More
4-Aminomethyl-2-chloropyridine, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 438890010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Aminomethyl-2-chloropyridine | |
94.0 | |
Authentic | |
Glass Bottle | |
1g | |
GGHCWJWUOSNCSK-UHFFFAOYSA-N | |
(2-chloropyridin-4-yl)methanamine | |
2794167 | |
95% |
144900-57-2 | |
100.0 | |
94% min. (GC) | |
C6H7ClN2 | |
2-chloropyridin-4-yl methanamine, 4-pyridinemethanamine, 2-chloro, 2-chloropyridin-4-yl methylamine, 4-aminomethyl-2-chloropyridine, 2-chloro-4-aminomethylpyridine, 2-chloro-4-pyridyl methylamine, 1-2-chloro-4-pyridinyl methanamine, 2-chloro-4-pyridinemethanamine, 1-2-chloropyridin-4-yl methanamine, c-2-chloro-pyridin-4-yl-methylamine | |
C1=CN=C(C=C1CN)Cl | |
142.59 | |
142.59 |
Chemical Identifiers
144900-57-2 | |
142.59 | |
2-chloropyridin-4-yl methanamine, 4-pyridinemethanamine, 2-chloro, 2-chloropyridin-4-yl methylamine, 4-aminomethyl-2-chloropyridine, 2-chloro-4-aminomethylpyridine, 2-chloro-4-pyridyl methylamine, 1-2-chloro-4-pyridinyl methanamine, 2-chloro-4-pyridinemethanamine, 1-2-chloropyridin-4-yl methanamine, c-2-chloro-pyridin-4-yl-methylamine | |
(2-chloropyridin-4-yl)methanamine |
C6H7ClN2 | |
GGHCWJWUOSNCSK-UHFFFAOYSA-N | |
2794167 | |
C1=CN=C(C=C1CN)Cl |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO â Research Use Only