Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Aminomethyl-1-(n-butyl)piperidine, 97%

$91.91 - $317.09

Chemical Identifiers

Chemical Name or Material 4-Aminomethyl-1-(n-butyl)piperidine
CAS 65017-57-4
Molecular Formula C10H22N2
Formula Weight 170.3
MDL Number MFCD08060490
Synonym 1-(n-Butyl)-4-piperidinemethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5094703

  SDS
alfa aesar™
H5094703
1g Each for $317.09

AAH50947MD

  SDS
alfa aesar™
H50947MD
250mg Each for $91.91
Description & Specifications

Specifications

Formula Weight 170.3
Assay 97%
Molecular Formula C10H22N2
MDL Number MFCD08060490
Quantity 1g
Chemical Name or Material 4-Aminomethyl-1-(n-butyl)piperidine
CAS 65017-57-4
Synonym 1-(n-Butyl)-4-piperidinemethylamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.