Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Aminoindole, 98%

$56.50 - $182.32

Chemical Identifiers

Chemical Name or Material 4-Aminoindole
CAS 5192-23-4
Molecular Formula C8H8N2
Formula Weight 132.17
MDL Number MFCD01076559
Beilstein 114919
Synonym 4-Indolamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL17792MC

  SDS
alfa aesar™
L17792MC
100mg Each for $56.50

AAL17792ME

  SDS
alfa aesar™
L17792ME
500mg Each for $182.32
Description & Specifications

Specifications

Formula Weight 132.17
Chemical Name or Material 4-Aminoindole
Synonym 4-Indolamine
MDL Number MFCD01076559
Health Hazard 1 Danger
Recommended Storage −30° to −10°C
CAS 5192-23-4
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C8H8N2
Beilstein 114919
Melting Point 107° to 108°C
Quantity 100mg
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C