Please login to your online account to display your discounted pricing

4-Aminoindole, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Aminoindole
Name Note 97%
CAS 5192-23-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H8N2
Formula Weight 132.16
MDL Number MFCD01076559
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC368615000

  SDS
fisher scientific acros
AC368615000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 132.16
Color Greenish-gray to tan
Quantity 500mg
Beilstein 22, IV, 4296
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Chemical Name or Material 4-Aminoindole
Name Note 97%
Assay Percent Range 97%
Molecular Formula C8H8N2
MDL Number MFCD01076559
CAS 5192-23-4
Synonym 4-Indolamine
Packaging Glass bottle